Gene Transcripts

Gene Symbol
ANGEL2
Entrez Gene ID
90806
Gene Name
angel homolog 2
Gene Aliases
Ccr4d, KIAA0759L
Chromosome Location
Chr. 1: 212992182 - 213015875 on Build GRCh38
UniGene ID
Hs.157078
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300753.1 NP_001287682.1 5 1044
NM_001300755.1 NP_001287684.1 5 1142
NM_001300757.1 NP_001287686.1 4 797
NM_001300758.1 NP_001287687.1 4 793
NM_144567.4 NP_653168.2 5 1119
NR_125333.1 4 793
XM_005273344.1 XP_005273401.1 5 1140
XM_005273345.1 XP_005273402.1 5 1186
XM_005273346.2 XP_005273403.1 5 939
XM_005273347.3 XP_005273404.1 4 811
XM_017002774.1 XP_016858263.1 5 1068
XM_017002775.1 XP_016858264.1 6 1367
XM_017002776.1 XP_016858265.1 6 1391
XM_017002777.1 XP_016858266.1 6 1437
XM_017002778.1 XP_016858267.1 6 1180
GenBank mRNA AF510741.1 - 4 748
AK295116.1 - 5 1048
AK299208.1 - 5 1112
AK299365.1 - 4 703
AK310946.1 - 3 285
AK311276.1 - 4 702
AL079275.1 - 5 1077
BC047469.1 - 4 709 5274835

View Full Product Details