Gene Transcripts

Gene Symbol
ISX
Entrez Gene ID
91464
Gene Name
intestine specific homeobox
Gene Aliases
Pix-1, RAXLX
Chromosome Location
Chr. 22: 35066136 - 35087387 on Build GRCh38
UniGene ID
Hs.567637
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303508.1 NP_001290437.1 2 754
GenBank mRNA CR749372.1 - 2 755

View Full Product Details