Gene Transcripts

Gene Symbol
MFSD4B
Entrez Gene ID
91749
Gene Name
major facilitator superfamily domain containing 4B
Gene Aliases
KIAA1919, NaGLT1
Chromosome Location
Chr. 6: 111259279 - 111280005 on Build GRCh38
UniGene ID
Hs.400572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153369.2 NP_699200.2 4 982
XM_017011483.1 XP_016866972.1 3 865
GenBank mRNA AB067506.1 - 1 217
AK093136.1 - 4 941
AK292735.1 - 4 911
AK315016.1 - 4 904
BC036115.1 - 4 891 5298999

View Full Product Details