Gene Transcripts

Gene Symbol
DIAPH1
Entrez Gene ID
1729
Gene Name
diaphanous related formin 1
Gene Aliases
DFNA1, DIA1, DRF1, LFHL1, SCBMS, hDIA1
Chromosome Location
Chr. 5: 141515021 - 141619055 on Build GRCh38
UniGene ID
Hs.529451
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079812.2 NP_001073280.1 10 1191
NM_001314007.1 NP_001300936.1 11 1218
NM_005219.4 NP_005210.3 11 1218
XM_011537572.2 XP_011535874.1 10 1159
XM_011537573.2 XP_011535875.1 11 1026
GenBank mRNA AB209482.1 - 11 1159
AF051782.1 - 10 1050
AK023345.1 - 4 401
AK304291.1 - 10 997
AK307822.1 - 7 650
AY363395.1 - 11 1077
BC117257.1 - 10 1068 40125808
BC143413.1 - 11 1095 9051921
BC143414.1 - 11 1095
DQ067453.1 - 10 1065

View Full Product Details