Gene Transcripts

Gene Symbol
CENPBD1
Entrez Gene ID
92806
Gene Name
CENPB DNA-binding domain containing 1
Gene Aliases
Chromosome Location
Chr. 16: 89969775 - 89972832 on Build GRCh38
UniGene ID
Hs.513832
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145039.3 NP_659476.2 1 1110
GenBank mRNA AK056131.1 - 1 749
AK315352.1 - 1 743
CD518450.1 - 1 148

View Full Product Details