Gene Transcripts

Gene Symbol
HAUS1
Entrez Gene ID
115106
Gene Name
HAUS augmin like complex subunit 1
Gene Aliases
CCDC5, HEI-C, HEIC, HsT1461
Chromosome Location
Chr. 18: 46104332 - 46128333 on Build GRCh38
UniGene ID
Hs.436617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138443.3 NP_612452.1 5 631
NR_026978.1 5 671
GenBank mRNA AK024747.1 - 2 94
AK097403.1 - 5 612
AK315605.1 - 5 577
AY360137.1 - 5 631
BC005958.1 - 5 607 4047707
BC014003.2 - 5 604 3844806

View Full Product Details