Gene Transcripts

Gene Symbol
ZMYND19
Entrez Gene ID
116225
Gene Name
zinc finger MYND-type containing 19
Gene Aliases
MIZIP
Chromosome Location
Chr. 9: 137582079 - 137590485 on Build GRCh38
UniGene ID
Hs.128096
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138462.2 NP_612471.1 3 380
XM_005266052.4 XP_005266109.1 3 809
XM_017014255.1 XP_016869744.1 3 813
GenBank mRNA AJ298882.2 - 3 388
AX766496.1 - 3 388
BC012948.1 - 3 343 4519078
BU147462.1 - 380

View Full Product Details