Gene Transcripts

Gene Symbol
ACSM1
Entrez Gene ID
116285
Gene Name
acyl-CoA synthetase medium-chain family member 1
Gene Aliases
BUCS1, MACS1
Chromosome Location
Chr. 16: 20623235 - 20697788 on Build GRCh38
UniGene ID
Hs.306812
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318890.1 NP_001305819.1 2 342
NM_052956.2 NP_443188.2 1 206
NR_134918.1 1 206
XM_006721016.2 XP_006721079.1 1 185
XM_006721017.2 XP_006721080.1 1 185
XM_011545729.2 XP_011544031.1 1 185
XM_011545730.2 XP_011544032.1 1 185
XM_017022914.1 XP_016878403.1 1 185
GenBank mRNA AB059429.1 - 1 206
AK314282.1 - 2 298
BC125177.1 - 1 206
BC125178.1 - 1 206 40121956
CN360958.1 - 3 377

View Full Product Details