Gene Transcripts

Gene Symbol
HSF5
Entrez Gene ID
124535
Gene Name
heat shock transcription factor 5
Gene Aliases
HSF 5, HSTF 5
Chromosome Location
Chr. 17: 58420167 - 58488401 on Build GRCh38
UniGene ID
Hs.380061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080439.2 NP_001073908.2 4 1167
XM_011524283.1 XP_011522585.1 4 1180
GenBank mRNA BC033017.1 - 4 1131
BC033020.1 - 4 1154 4823233
BC115385.1 - 5 743 40038134
BC115386.1 - 5 739

View Full Product Details