Gene Transcripts

Gene Symbol
C5orf47
Entrez Gene ID
133491
Gene Name
chromosome 5 open reading frame 47
Gene Aliases
Chromosome Location
Chr. 5: 173989159 - 174007524 on Build GRCh38
UniGene ID
Hs.131469
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144954.1 NP_001138426.1 2 456
XM_011534431.1 XP_011532733.1 2 450
XM_017009028.1 XP_016864517.1 2 450
GenBank mRNA BC034971.1 - 2 422
BC047225.1 - 2 411
BC053953.1 - 2 411
BC057842.1 - 2 397
BC066964.1 - 2 418
BC092489.1 - 2 445

View Full Product Details