Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004420.2 | NP_004411.2 | 5 | 778 | |
XM_011519932.2 | XP_011518234.1 | 5 | 1805 | ||
XM_011519933.2 | XP_011518235.1 | 5 | 835 | ||
GenBank mRNA | BC038231.1 | - | 4 | 781 | |
BC045110.1 | - | 4 | 651 | 5478677 | |
U27193.1 | - | 5 | 784 |