Gene Transcripts
- Gene Symbol
- SPTY2D1
- Entrez Gene ID
- 144108
- Gene Name
- SPT2 chromatin protein domain containing 1
- Gene Aliases
- Spt2
- Chromosome Location
- Chr. 11: 18606401 - 18634823 on Build GRCh38
- UniGene ID
- Hs.738650
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_194285.2 |
NP_919261.2
|
5, 6 |
2188 |
|
XM_011519919.2 |
XP_011518221.1
|
5, 6 |
2046 |
|
GenBank mRNA |
AK096694.1 |
- |
1, 2 |
92 |
|
AL834393.1 |
- |
2, 3 |
238 |
|
BC036844.1 |
- |
3, 4 |
808 |
|
BX538046.1 |
- |
3, 4 |
567 |
|
BX641102.1 |
- |
5, 6 |
2089 |
|
BX647798.1 |
- |
5, 6 |
2188 |
|
BX648114.1 |
- |
3, 4 |
1767 |
|
CR749626.1 |
- |
3, 4 |
1631 |
|
View Full Product Details