Gene Transcripts

Gene Symbol
CPNE8
Entrez Gene ID
144402
Gene Name
copine 8
Gene Aliases
Chromosome Location
Chr. 12: 38652200 - 38908593 on Build GRCh38
UniGene ID
Hs.40910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153634.2 NP_705898.1 10, 11 800
XM_011537951.2 XP_011536253.1 10, 11 3775
XM_011537952.2 XP_011536254.1 10, 11 3775
XM_017018851.1 XP_016874340.1 11, 12 1773
XM_017018852.1 XP_016874341.1 10, 11 746
GenBank mRNA AK098593.1 - 10, 11 730
AK307908.1 - 10, 11 773
AY177785.1 - 10, 11 716
BC068040.1 - 10, 11 764
BC110577.1 - 10, 11 791 40007743
BC110578.1 - 10, 11 791 40007744
BX538055.1 - 10, 11 814

View Full Product Details