Gene Transcripts

Gene Symbol
GCNT7
Entrez Gene ID
140687
Gene Name
glucosaminyl (N-acetyl) transferase family member 7
Gene Aliases
C20orf105, dJ1153D9.2, gcnt
Chromosome Location
Chr. 20: 56491492 - 56525925 on Build GRCh38
UniGene ID
Hs.655916
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080615.1 NP_542182.1 5 1457
GenBank mRNA AK131203.1 - 5 1457

View Full Product Details