Gene Transcripts

Gene Symbol
OTUD7A
Entrez Gene ID
161725
Gene Name
OTU deubiquitinase 7A
Gene Aliases
C15orf16, C16ORF15, CEZANNE2, OTUD7
Chromosome Location
Chr. 15: 31483126 - 31655339 on Build GRCh38
UniGene ID
Hs.355236
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130901.1 NP_570971.1 5 775
GenBank mRNA AJ430383.1 - 5 775
BC035668.1 - 7 851

View Full Product Details