Gene Transcripts
- Gene Symbol
- TRPV3
- Entrez Gene ID
- 162514
- Gene Name
- transient receptor potential cation channel subfamily V member 3
- Gene Aliases
- FNEPPK2, OLMS, VRL3
- Chromosome Location
- Chr. 17: 3510502 - 3557995 on Build GRCh38
- UniGene ID
- Hs.446255
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001258205.1 |
NP_001245134.1
|
3, 4 |
552 |
|
NM_145068.3 |
NP_659505.1
|
3, 4 |
552 |
|
GenBank mRNA |
AF514998.1 |
- |
3, 4 |
294 |
|
AJ487035.2 |
- |
3, 4 |
552 |
|
AY118267.1 |
- |
3, 4 |
362 |
|
AY118268.1 |
- |
3, 4 |
362 |
|
BC104866.1 |
- |
3, 4 |
324 |
8143869 |
BC104868.1 |
- |
3, 4 |
324 |
8143871 |
BC143294.1 |
- |
4, 5 |
493 |
9051797 |
BC143295.1 |
- |
4, 5 |
493 |
|
BC143299.1 |
- |
4, 5 |
493 |
9051803 |
BC143330.1 |
- |
3, 4 |
324 |
9051835 |
BC143333.1 |
- |
4, 5 |
413 |
|
EU295940.1 |
- |
2, 3 |
230 |
|
View Full Product Details