Gene Transcripts

Gene Symbol
IFNLR1
Entrez Gene ID
163702
Gene Name
interferon lambda receptor 1
Gene Aliases
CRF2/12, IFNLR, IL-28R1, IL28RA, LICR2
Chromosome Location
Chr. 1: 24154156 - 24187275 on Build GRCh38
UniGene ID
Hs.221375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170743.3 NP_734464.1 5 634
NM_173064.2 NP_775087.1 5 634
NM_173065.2 NP_775088.1 5 634
XM_006710394.3 XP_006710457.2 4 698
XM_017000479.1 XP_016855968.1 4 663
GenBank mRNA AF439325.1 - 5 607
AJ534330.1 - 5 620
AJ534331.1 - 5 620
AK160364.1 - 5 610
AY129151.1 - 5 607
AY129152.1 - 5 607
BC101407.1 - 4 496
BC101408.1 - 4 496
BC101975.1 - 5 620
BC139722.1 - 5 620
BC140872.1 - 5 683 9021740
JN591376.1 - 5 607

View Full Product Details