Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004097.2 | NP_004088.2 | 3 | 1117 | |
XM_011532697.2 | XP_011530999.1 | 3 | 6137 | ||
GenBank mRNA | AK092368.1 | - | 1 | 649 | |
BC037242.1 | - | 3 | 984 | ||
BC045762.1 | - | 3 | 1117 | 5260039 | |
X68879.1 | - | 3 | 328 |