Gene Transcripts

Gene Symbol
IBA57
Entrez Gene ID
200205
Gene Name
IBA57 homolog, iron-sulfur cluster assembly
Gene Aliases
C1orf69, MMDS3, SPG74
Chromosome Location
Chr. 1: 228165808 - 228182257 on Build GRCh38
UniGene ID
Hs.237017
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010867.3 NP_001010867.1 1 307
GenBank mRNA BI820993.1 - 1 48
CN285775.1 - 1 387
HY081013.1 - 1 307

View Full Product Details