Gene Transcripts

Gene Symbol
TEX37
Entrez Gene ID
200523
Gene Name
testis expressed 37
Gene Aliases
C2orf51, TSC21
Chromosome Location
Chr. 2: 88524623 - 88529585 on Build GRCh38
UniGene ID
Hs.132104
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152670.2 NP_689883.1 3 206
XM_005264188.3 XP_005264245.3 3 181
XM_011532691.1 XP_011530993.1 4 300
XM_011532692.1 XP_011530994.1 3 234
GenBank mRNA AK058098.1 - 3 206
BC029522.1 - 3 173 5297675

View Full Product Details