Gene Transcripts

Gene Symbol
TBATA
Entrez Gene ID
219793
Gene Name
thymus, brain and testes associated
Gene Aliases
C10orf27, SPATIAL
Chromosome Location
Chr. 10: 70771237 - 70785419 on Build GRCh38
UniGene ID
Hs.386698
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318241.1 NP_001305170.1 10 1353
NM_001318242.1 NP_001305171.1 10 1417
NM_001318243.1 NP_001305172.1 10 1347
NM_152710.3 NP_689923.3 10 1350
NR_134531.1 9 1268
NR_134532.1 10 1276
NR_134533.1 6 886
NR_134534.1 5 810
XM_017015845.1 XP_016871334.1 11 1523
XM_017015846.1 XP_016871335.1 11 1523
XM_017015847.1 XP_016871336.1 11 1523
XM_017015848.1 XP_016871337.1 11 1477
XM_017015849.1 XP_016871338.1 11 1523
XM_017015853.1 XP_016871342.1 11 1428
XM_017015854.1 XP_016871343.1 11 1477
XM_017015855.1 XP_016871344.1 11 1477
XM_017015856.1 XP_016871345.1 11 1428
XM_017015857.1 XP_016871346.1 11 1428
XM_017015860.1 XP_016871349.1 9 1201
GenBank mRNA AK057382.1 - 10 1332
AK124147.1 - 1 1425
BC137543.1 - 10 1384 9021543
BC137544.1 - 10 1318 9021544
BC139728.1 - 9 1217 40036949
BC144692.1 - 10 1315 9053226
BC144694.1 - 10 1321 9053228
DB512647.2 - 4 322

View Full Product Details