Gene Transcripts

Gene Symbol
OR4D11
Entrez Gene ID
219986
Gene Name
olfactory receptor family 4 subfamily D member 11
Gene Aliases
OR4D11P
Chromosome Location
Chr. 11: 59503576 - 59504511 on Build GRCh38
UniGene ID
Hs.553651
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004706.1 NP_001004706.1 1 577

View Full Product Details