Gene Transcripts

Gene Symbol
GAS2L2
Entrez Gene ID
246176
Gene Name
growth arrest specific 2 like 2
Gene Aliases
GAR17
Chromosome Location
Chr. 17: 35744511 - 35752878 on Build GRCh38
UniGene ID
Hs.591194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139285.3 NP_644814.1 6 1619
GenBank mRNA AF508784.1 - 7 1572
AF508785.1 - 6 1619

View Full Product Details