Gene Transcripts

Gene Symbol
SLC17A8
Entrez Gene ID
246213
Gene Name
solute carrier family 17 member 8
Gene Aliases
DFNA25, VGLUT3
Chromosome Location
Chr. 12: 100357079 - 100422059 on Build GRCh38
UniGene ID
Hs.116871
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139319.2 NP_647480.1 7, 8 1209
GenBank mRNA AJ459241.1 - 7, 8 1072
AK128319.1 - 7, 8 1209

View Full Product Details