Gene Transcripts

Gene Symbol
MFSD8
Entrez Gene ID
256471
Gene Name
major facilitator superfamily domain containing 8
Gene Aliases
CCMD, CLN7
Chromosome Location
Chr. 4: 127917805 - 127965984 on Build GRCh38
UniGene ID
Hs.480701
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152778.2 NP_689991.1 5 583
XM_005262893.1 XP_005262950.1 4 497
XM_005262896.1 XP_005262953.1 4 497
XM_005262897.2 XP_005262954.1 4 497
XM_005262898.2 XP_005262955.1 4 497
XM_011531830.1 XP_011530132.1 4 497
XM_011531831.1 XP_011530133.1 4 497
XM_011531832.1 XP_011530134.1 4 497
XM_017007989.1 XP_016863478.1 4 497
GenBank mRNA AK074564.1 - 4 432
AK294184.1 - 4 400
AK294439.1 - 4 429
AK294527.1 - 4 406
AK294957.1 - 4 425
AK315596.1 - 4 429
BC029503.1 - 5 548 5272664

View Full Product Details