Gene Transcripts
- Gene Symbol
- MFSD8
- Entrez Gene ID
- 256471
- Gene Name
- major facilitator superfamily domain containing 8
- Gene Aliases
- CCMD, CLN7
- Chromosome Location
- Chr. 4: 127917805 - 127965984 on Build GRCh38
- UniGene ID
- Hs.480701
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152778.2 |
NP_689991.1
|
8, 9 |
926 |
|
XM_005262893.1 |
XP_005262950.1
|
7, 8 |
840 |
|
XM_005262896.1 |
XP_005262953.1
|
6, 7 |
693 |
|
XM_005262897.2 |
XP_005262954.1
|
5, 6 |
639 |
|
XM_005262898.2 |
XP_005262955.1
|
7, 8 |
840 |
|
XM_011531830.1 |
XP_011530132.1
|
6, 7 |
726 |
|
XM_011531831.1 |
XP_011530133.1
|
4, 5 |
525 |
|
XM_011531832.1 |
XP_011530134.1
|
6, 7 |
726 |
|
XM_017007989.1 |
XP_016863478.1
|
5, 6 |
639 |
|
GenBank mRNA |
AK074564.1 |
- |
7, 8 |
775 |
|
AK294184.1 |
- |
5, 6 |
573 |
|
AK294439.1 |
- |
6, 7 |
658 |
|
AK294527.1 |
- |
6, 7 |
604 |
|
AK294957.1 |
- |
7, 8 |
768 |
|
AK315596.1 |
- |
7, 8 |
772 |
|
BC029503.1 |
- |
8, 9 |
891 |
5272664 |
View Full Product Details