Gene Transcripts
- Gene Symbol
- C9orf43
- Entrez Gene ID
- 257169
- Gene Name
- chromosome 9 open reading frame 43
- Gene Aliases
- Chromosome Location
- Chr. 9: 113410644 - 113429684 on Build GRCh38
- UniGene ID
- Hs.632691
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278629.1 |
NP_001265558.1
|
7, 8 |
1058 |
|
NM_001278630.1 |
NP_001265559.1
|
7, 8 |
915 |
|
NM_152786.2 |
NP_689999.1
|
7, 8 |
1131 |
|
XM_005251885.3 |
XP_005251942.1
|
5, 6 |
437 |
|
XM_011518477.1 |
XP_011516779.1
|
7, 8 |
1058 |
|
XM_011518478.1 |
XP_011516780.1
|
7, 8 |
1058 |
|
XM_011518479.2 |
XP_011516781.1
|
5, 6 |
713 |
|
GenBank mRNA |
AK301872.1 |
- |
7, 8 |
915 |
|
AK313867.1 |
- |
7, 8 |
1047 |
|
BC026884.1 |
- |
7, 8 |
1097 |
3455891 |
BC033037.2 |
- |
7, 8 |
1016 |
4827930 |
View Full Product Details