Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190707.1 | NP_001177636.1 | 20 | 2398 | |
NM_147129.4 | NP_667340.2 | 20 | 2301 | ||
NR_033815.2 | 20 | 2587 | |||
NR_135622.1 | 20 | 2297 | |||
XM_005265025.1 | XP_005265082.1 | 9 | 950 | ||
XM_006713091.2 | XP_006713154.1 | 20 | 2571 | ||
XM_017006120.1 | XP_016861609.1 | 20 | 2281 | ||
GenBank mRNA | AB107015.1 | - | 20 | 2289 | |
AK074118.1 | - | 17 | 2551 | ||
AK092455.1 | - | 5 | 678 | ||
AK093844.1 | - | 9 | 950 | ||
AK126505.1 | - | 4 | 909 | ||
AK131270.1 | - | 20 | 2395 | ||
AK131273.1 | - | 17 | 1863 | ||
AL832132.1 | - | 20 | 2578 | ||
AL832203.1 | - | 17 | 4674 | ||
BC042906.1 | - | 12 | 1331 | ||
BC061883.1 | - | 12 | 1331 | ||
BC075825.1 | - | 12 | 1331 | ||
BC109233.1 | - | 4 | 753 | ||
CR627258.1 | - | 17 | 4642 |