Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181655.3 | NP_858041.2 | 3 | 1287 | |
NM_181656.4 | NP_858042.2 | 3 | 1430 | ||
GenBank mRNA | AK026583.1 | - | 3 | 1368 | |
BC021702.1 | - | 2 | 1046 | ||
BC036760.1 | - | 3 | 1159 | 5587712 |