Gene Transcripts

Gene Symbol
RHOBTB1
Entrez Gene ID
9886
Gene Name
Rho related BTB domain containing 1
Gene Aliases
Chromosome Location
Chr. 10: 60869438 - 61001486 on Build GRCh38
UniGene ID
Hs.737374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242359.1 NP_001229288.1 7 1829
NM_014836.4 NP_055651.1 6 1757
NR_024554.2 7 1631
NR_024555.1 7 1595
XM_006718084.1 XP_006718147.1 6 1565
XM_006718085.1 XP_006718148.1 6 1592
XM_011540424.1 XP_011538726.1 7 1623
XM_011540425.1 XP_011538727.1 7 1623
XM_011540426.1 XP_011538728.1 7 1623
XM_011540428.1 XP_011538730.1 7 1623
XM_017017000.1 XP_016872489.1 6 1812
XM_017017001.1 XP_016872490.1 6 1808
XM_017017002.1 XP_016872491.1 6 1808
XM_017017003.1 XP_016872492.1 8 1698
XM_017017004.1 XP_016872493.1 8 1700
XM_017017005.1 XP_016872494.1 7 1605
XM_017017006.1 XP_016872495.1 7 1654
XM_017017007.1 XP_016872496.1 5 1704
XM_017017008.1 XP_016872497.1 7 1865
GenBank mRNA AB018283.3 - 6 1648
AK075129.1 - 7 1595
AK307630.1 - 5 1327
BC032848.1 - 7 1806 4830471
BC041791.1 - 6 1548 5297756

View Full Product Details