Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142547.1 | NP_001136019.1 | 7 | 710 | |
NM_173659.3 | NP_775930.2 | 8 | 755 | ||
GenBank mRNA | AK291888.1 | - | 8 | 756 | |
BC032135.2 | - | 8 | 729 | 4649090 | |
BC062362.1 | - | 7 | 686 | 5454417 | |
BC065741.1 | - | 8 | 757 | 4091801 |