Gene Transcripts

Gene Symbol
ATG9B
Entrez Gene ID
285973
Gene Name
autophagy related 9B
Gene Aliases
APG9L2, NOS3AS, SONE
Chromosome Location
Chr. 7: 151012209 - 151024499 on Build GRCh38
UniGene ID
Hs.707300
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317056.1 NP_001303985.1 1 384
NR_133652.1 1 384
XM_011516065.1 XP_011514367.1 1 384
XM_011516066.2 XP_011514368.1 1 384
GenBank mRNA AK096734.1 - 1 384
AY515311.1 - 1 384

View Full Product Details