Gene Transcripts
- Gene Symbol
- C8orf31
- Entrez Gene ID
- 286122
- Gene Name
- chromosome 8 open reading frame 31
- Gene Aliases
- Chromosome Location
- Chr. 8: 143038983 - 143054303 on Build GRCh38
- UniGene ID
- Hs.660382
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173687.3 |
NP_775958.1
|
2, 3 |
495 |
|
GenBank mRNA |
AK094450.1 |
- |
2, 3 |
443 |
|
View Full Product Details