Gene Transcripts

Gene Symbol
C8orf31
Entrez Gene ID
286122
Gene Name
chromosome 8 open reading frame 31
Gene Aliases
Chromosome Location
Chr. 8: 143038983 - 143054303 on Build GRCh38
UniGene ID
Hs.660382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173687.3 NP_775958.1 3 570
NR_126398.1 3 555
NR_126399.1 2 425
XM_017013344.1 XP_016868833.1 3 781
XM_017013345.1 XP_016868834.1 2 176
XM_017013346.1 XP_016868835.1 2 176
GenBank mRNA AK094450.1 - 3 518
BC063497.1 - 2 336
BC073830.1 - 3 503
BC120993.2 - 2 360
DB260703.1 - 2 425

View Full Product Details