Gene Transcripts

Gene Symbol
RAB37
Entrez Gene ID
326624
Gene Name
RAB37, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 17: 74671117 - 74747335 on Build GRCh38
UniGene ID
Hs.351413
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006638.2 NP_001006639.1 9 669
NM_001163989.1 NP_001157461.1 9 818
NM_001163990.1 NP_001157462.1 8 558
NM_175738.4 NP_783865.1 9 1062
XM_005257288.4 XP_005257345.1 9 687
GenBank mRNA AK054846.1 - 6 1797
AK057069.1 - 3 213
AK098068.1 - 9 669
AK127073.1 - 8 1822
AK290202.1 - 9 1038
AK296062.1 - 9 657
AK296172.1 - 8 521
AK303442.1 - 9 721
BC016615.1 - 9 615 4520191
BC040547.1 - 9 1048 5287038

View Full Product Details