Gene Transcripts

Gene Symbol
KRTAP13-3
Entrez Gene ID
337960
Gene Name
keratin associated protein 13-3
Gene Aliases
KAP13.3
Chromosome Location
Chr. 21: 30425393 - 30425912 on Build GRCh38
UniGene ID
Hs.553687
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181622.1 NP_853653.1 1 453
GenBank mRNA AB096940.1 - 1 467
BC103965.2 - 1 453 40014483

View Full Product Details