Gene Transcripts

Gene Symbol
TEX33
Entrez Gene ID
339669
Gene Name
testis expressed 33
Gene Aliases
C22orf33, EAN57, cE81G9.2
Chromosome Location
Chr. 22: 36991119 - 37007864 on Build GRCh38
UniGene ID
Hs.124502
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163857.1 NP_001157329.1 4 733
NM_178552.3 NP_848647.1 4 714
XM_011530165.2 XP_011528467.1 5 946
XM_011530166.1 XP_011528468.1 4 689
GenBank mRNA BC042635.1 - 4 676
CR456416.1 - 4 699

View Full Product Details