Gene Transcripts

Gene Symbol
SPOPL
Entrez Gene ID
339745
Gene Name
speckle type BTB/POZ protein like
Gene Aliases
BTBD33
Chromosome Location
Chr. 2: 138501780 - 138573235 on Build GRCh38
UniGene ID
Hs.333297
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001664.2 NP_001001664.1 6 922
XM_005263655.4 XP_005263712.1 7 885
XM_005263656.4 XP_005263713.1 7 814
XM_005263657.4 XP_005263714.1 6 727
GenBank mRNA BC071613.1 - 6 901

View Full Product Details