Gene Transcripts

Gene Symbol
NHSL2
Entrez Gene ID
340527
Gene Name
NHS like 2
Gene Aliases
Chromosome Location
Chr. X: 71911073 - 72153286 on Build GRCh38
UniGene ID
Hs.397836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013627.2 NP_001013649.2 6 2402
XM_011530932.1 XP_011529234.1 7 2486
XM_011530933.1 XP_011529235.1 6 2354
XM_011530934.2 XP_011529236.1 6 7194
XM_017029473.1 XP_016884962.1 6 6164
XM_017029474.1 XP_016884963.1 6 2306
XM_017029475.1 XP_016884964.1 2 5549
XM_017029476.1 XP_016884965.1 2 1666
XM_017029477.1 XP_016884966.1 2 6580
GenBank mRNA BC033261.1 - 2 2566
BC136756.1 - 3 1775 9020746

View Full Product Details