Gene Transcripts

Gene Symbol
PLET1
Entrez Gene ID
349633
Gene Name
placenta expressed transcript 1
Gene Aliases
C11orf34
Chromosome Location
Chr. 11: 112248153 - 112260860 on Build GRCh38
UniGene ID
Hs.632097
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145024.1 NP_001138496.1 1 446
GenBank mRNA AY364431.1 - 1 175

View Full Product Details