Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021250.3 | NP_067073.1 | 5 | 641 | |
NM_181879.2 | NP_870994.1 | 5 | 641 | ||
NM_181985.3 | NP_871714.1 | 4 | 605 | ||
NM_181986.2 | NP_871715.1 | 4 | 605 | ||
GenBank mRNA | AF212842.1 | - | 2 | 355 | |
AF324830.1 | - | 5 | 641 | ||
AF499916.1 | - | 5 | 584 | ||
AF499917.1 | - | 4 | 540 | ||
AF499918.1 | - | 5 | 600 | ||
AF499919.1 | - | 4 | 557 | ||
EU915611.1 | - | 5 | 581 | ||
EU915612.1 | - | 4 | 545 |