Gene Transcripts

Gene Symbol
USP50
Entrez Gene ID
373509
Gene Name
ubiquitin specific peptidase 50
Gene Aliases
Chromosome Location
Chr. 15: 50500562 - 50546705 on Build GRCh38
UniGene ID
Hs.677758
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203494.4 NP_987090.2 4 740
GenBank mRNA AJ583818.2 - 4 575

View Full Product Details