Gene Transcripts

Gene Symbol
PTCHD3
Entrez Gene ID
374308
Gene Name
patched domain containing 3
Gene Aliases
PTR
Chromosome Location
Chr. 10: 27398188 - 27414368 on Build GRCh38
UniGene ID
Hs.631832
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034842.3 NP_001030014.2 2, 3 1337
GenBank mRNA BC107139.2 - 2, 3 1261
JF332167.1 - 2, 3 1219

View Full Product Details