Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198557.2 | NP_940959.1 | 2 | 333 | |
XM_017004052.1 | XP_016859541.1 | 2 | 790 | ||
GenBank mRNA | AK127552.1 | - | 2 | 322 | |
BC136410.1 | - | 2 | 233 | 9020398 | |
BC136411.1 | - | 2 | 233 | 9020399 | |
DA658209.1 | - | 2 | 311 |