Gene Transcripts

Gene Symbol
C1QL3
Entrez Gene ID
389941
Gene Name
complement C1q like 3
Gene Aliases
C1QTNF13, C1ql, CTRP13, K100
Chromosome Location
Chr. 10: 16513743 - 16522005 on Build GRCh38
UniGene ID
Hs.676792
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010908.1 NP_001010908.1 1, 2 1536
GenBank mRNA BC127716.1 - 2, 3 604
BC127717.1 - 2, 3 550 40127921

View Full Product Details