Gene Transcripts

Gene Symbol
OR5B2
Entrez Gene ID
390190
Gene Name
olfactory receptor family 5 subfamily B member 2
Gene Aliases
OR11-240, OST073
Chromosome Location
Chr. 11: 58422265 - 58423313 on Build GRCh38
UniGene ID
Hs.553753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005566.2 NP_001005566.1 1 652
GenBank mRNA BC136922.1 - 1 655

View Full Product Details