Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005253.3 | NP_005244.1 | 4 | 1650 | |
XM_006711976.2 | XP_006712039.1 | 4 | 1854 | ||
XM_006711977.3 | XP_006712040.1 | 4 | 2707 | ||
XM_017003737.1 | XP_016859226.1 | 4 | 2363 | ||
GenBank mRNA | AK055579.1 | - | 4 | 1704 | |
AK302622.1 | - | 4 | 804 | ||
AK315415.1 | - | 4 | 954 | ||
BC022791.1 | - | 4 | 1074 | 4637727 | |
BX647822.1 | - | 4 | 826 | ||
CR542262.1 | - | 4 | 841 | ||
X16706.1 | - | 4 | 844 |