Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002031.2 | NP_002022.1 | 4 | 1101 | |
XM_005266880.4 | XP_005266937.1 | 5 | 1173 | ||
XM_005266881.2 | XP_005266938.1 | 5 | 1090 | ||
XM_005266882.4 | XP_005266939.1 | 6 | 1402 | ||
XM_011535653.2 | XP_011533955.1 | 6 | 1405 | ||
XM_011535654.2 | XP_011533956.1 | 5 | 1187 | ||
XM_011535655.2 | XP_011533957.1 | 5 | 1184 | ||
XM_011535656.2 | XP_011533958.1 | 4 | 558 | ||
XM_017010645.1 | XP_016866134.1 | 5 | 1291 | ||
GenBank mRNA | AK302264.1 | - | 3 | 276 | |
AK313314.1 | - | 4 | 1181 | ||
BC012916.1 | - | 4 | 946 | 4515877 | |
U00803.1 | - | 4 | 1101 | ||
U22322.1 | - | 4 | 1063 |