Gene Transcripts

Gene Symbol
PIWIL3
Entrez Gene ID
440822
Gene Name
piwi like RNA-mediated gene silencing 3
Gene Aliases
HIWI3
Chromosome Location
Chr. 22: 24719034 - 24774720 on Build GRCh38
UniGene ID
Hs.448343
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008496.3 NP_001008496.2 15 2288
NM_001255975.1 NP_001242904.1 15 2261
NR_045648.1 16 2471
NR_045649.1 16 2344
GenBank mRNA AB079368.1 - 15 2284
AK302409.1 - 15 2261
AK315806.1 - 15 2261
BC144693.1 - 16 2101
BC144696.1 - 16 2228

View Full Product Details