Gene Transcripts
- Gene Symbol
- PTMA
- Entrez Gene ID
- 5757
- Gene Name
- prothymosin, alpha
- Gene Aliases
- TMSA
- Chromosome Location
- Chr. 2: 231708525 - 231713541 on Build GRCh38
- UniGene ID
- Hs.459927
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002823.4 |
NP_002814.3
|
2, 3 |
291 |
|
XM_011511519.1 |
XP_011509821.1
|
2, 3 |
1021 |
|
XM_011511521.1 |
XP_011509823.1
|
2, 3 |
920 |
|
GenBank mRNA |
AF348514.1 |
- |
2, 3 |
109 |
|
AF452640.1 |
- |
2, 3 |
591 |
|
AK223605.1 |
- |
2, 3 |
286 |
|
AK314106.1 |
- |
2, 3 |
286 |
|
AY169282.1 |
- |
2, 3 |
109 |
|
BC003510.1 |
- |
2, 3 |
274 |
3610808 |
BC022433.1 |
- |
2, 3 |
290 |
4277490 |
BC034921.1 |
- |
2, 3 |
282 |
4335961 |
BC051265.1 |
- |
2, 3 |
274 |
3832454 |
BC066905.1 |
- |
2, 3 |
212 |
4830132 |
BC070480.1 |
- |
2, 3 |
276 |
6205582 |
BC071647.1 |
- |
2, 3 |
269 |
5112610 |
BC071879.1 |
- |
2, 3 |
271 |
6425103 |
BC098420.1 |
- |
2, 3 |
276 |
5017164 |
BM839980.1 |
- |
2, 3 |
291 |
|
M14483.1 |
- |
2, 3 |
241 |
|
M14794.1 |
- |
2, 3 |
177 |
|
M26708.1 |
- |
2, 3 |
264 |
|
- Gene Symbol
- LOC441454
- Entrez Gene ID
- 441454
- Gene Name
- prothymosin, alpha pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 9: 96909075 - 96910455 on Build GRCh38
- UniGene ID
- Hs.653264
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028350.1 |
|
1 |
1110 |
|
- Gene Symbol
- LOC728026
- Entrez Gene ID
- 728026
- Gene Name
- prothymosin alpha-like
- Gene Aliases
- Chromosome Location
- Chr. 9: 94347222 - 94348219 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details